missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ G2 Dialysis Cassettes, 10K MWCO

Important Note: These G2 Dialysis Cassettes will be discontinued on June 1, 2023 and replaced by G3 Dialysis Cassettes, a new generation of cassettes. Consider switching to G3 Dialysis Cassettes. Remove salts and contaminants while maximizing sample recovery with these second-generation, self-floating protein dialysis cassettes.

£147.00 - £257.00

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 10 kDa
View More Specs

Products 10
Product Code Brand Description Sample Volume (Metric) Price Quantity & Availability  
Product Code Brand Description Sample Volume (Metric) Price Quantity & Availability  
10401955
View Documents
Thermo Scientific™
87729
G2 Dialysis Cassettes 0.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10625655
View Documents
Thermo Scientific™
87730
G2 Dialysis Cassettes 3 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10717975
View Documents
Thermo Scientific™
87731
G2 Dialysis Cassettes 15 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10718165
View Documents
Thermo Scientific™
87732
G2 Dialysis Cassettes 30 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10318694
View Documents
Thermo Scientific™
87733
G2 Dialysis Cassettes 70 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11388271
View Documents
Thermo Scientific™
88250
G2 Dialysis Cassettes (gamma-irradiated) 0.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11582541
View Documents
Thermo Scientific™
88251
G2 Dialysis Cassettes (gamma-irradiated) 3 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11592541
View Documents
Thermo Scientific™
88252
G2 Dialysis Cassettes (gamma-irradiated) 15 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11398271
View Documents
Thermo Scientific™
88253
G2 Dialysis Cassettes (gamma-irradiated) 30 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11308281
View Documents
Thermo Scientific™
88254
G2 Dialysis Cassettes (gamma-irradiated) 70 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO) provide maximum convenience, flexibility, and performance for sample dialysis. They allow samples to be loaded with a syringe or pipette through the unique sample port. The sample is easily recovered through either the pipette or syringe sample ports. The Slide-A-Lyzer Cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems.

Slide-A-Lyzer G2 Dialysis Cassettes allow samples to be directly injected with a syringe or pipette through the unique sample port. The cassette design allows air to be removed from inside the cassette to maximize the surface area-to-volume ratio, providing better dialysis performance than dialysis tubing and tube-based systems. The sample is easily recovered through either the pipette or syringe sample ports.

The 10K molecular-weight cutoff (MWCO) cassettes use a low-binding regenerated cellulose that is rated to completely retain proteins and other macromolecules larger than 10,000 Daltons while allowing removal of buffer salts and small contaminants. Membranes and cassettes are manufactured using clean-room conditions to ensure that are contaminant-free.

Features of Slide-A-Lyzer G2 Dialysis Cassettes (10K MWCO):
Easy-loading—pipette-accessible for easy sample loading and retrieval
Self-floating—integrated air chambers eliminate the need for float buoys
Sturdy construction—ensures the highest possible sample integrity and protection
Superior design—thoroughly researched and tested to provide fast and consistent dialysis with maximum sample recovery
Versatile—ideal for removing low molecular-weight contaminants, performing buffer exchange and desalting
Multiple sizes—five cassette capacities to optimally match 0.5 to 70 mL sample volumes
Gamma-irradiated option—gamma-irradiated cassettes are available for applications requiring conditions that minimize the risk of sample contamination

Gamma-irradiated (sterile) dialysis cassettes
Gamma-irradiated Slide-A-Lyzer G2 Dialysis Cassettes are ideal for researchers culturing cells and microorganisms; purifying viruses, DNA, and RNA; or performing sample preparation for other applications requiring conditions that minimize the risk of sample contamination. The gamma-irradiated cassettes are treated with 20–36 kGy of gamma radiation, a dosage that has been determined to sterilize the units based on both trypticase soy broth medium for identifying total growth and fluid thioglycollate medium for identifying anaerobic growth. However, a confirmation of sterility is not provided for individual lots, as no further sterility testing is conducted post irradiation.

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: 21%
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.25 to 0.75 mL
• 3 mL = 1 to 3 mL
• 15 mL = 5 to 15 mL
• 30 mL = 10 to 30 mL
• 70 mL = 25 to 70 mL

Specifications

Specifications

Store at room temperature
Cellulose Membrane, Dialysis
10 kDa
Buffer Exchange, Protein
Cassette
Slide-A-Lyzer™
Videos
SDS
Documents

Documents

Product Certifications