Learn More
Thermo Scientific™ Float Buoys, for 0.1 mL Slide-A-Lyzer™ MINI Dialysis Devices
Thermo Scientific Slide-A-Lyzer MINI Dialysis Device Float buoys are low density polyethylene (LPDE) rafts designed to securely hold up to 25 Slide-A-Lyzer MINI Dialysis Devices (0.1 mL only) to permit array dialysis.
Brand: Thermo Scientific™ 69588
31.20 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Description
Float Buoys for Slide-A-Lyzer™ MINI Dialysis Devices contain inert, low-density polyethylene (LPDE) construction that is compatible with most aqueous buffer conditions, and the floats can be washed and reused. These Float Buoys enable users to dialyze a single sample (tube) or multiple samples (25 array). Their easy-to-read column and row labels make it simple to keep track of the identity of different samples.
Float Buoys for Slide-A-Lyzer MINI Dialysis Devices are intended to hold Slide-A-Lyzer MINI Dialysis Devices (0.1 mL size only), whose bottom surfaces are a dialysis interface composed of a cellulose membrane barrier. The float holds the devices so that the bottoms contact the solution in which they are floated for dialysis. LPDE material is not suitable for use at temperatures greater than 50°C and are not designed for use as water bath floats.Specifications
Store at room temperature. | |
MINI Dialysis Devices | |
Slide-A-Lyzer™ |
Cellulose Membrane, Dialysis | |
0.1 mL | |
4 Buoys |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.