Learn More
Epredia™ Slide Baskets and Hangers for Epredia™ ClearVue™ Coverslipper
Use these slide baskets and hangers with the Epredia™ ClearVue™ Coverslipper to make it compatible with any automated slide stainer.
Brand:  Epredia™ A79210065
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12638196
  • £125.00 / Pack of 5
Estimated Shipment: 22-07-2024
to see stock.
Description
Description
  • Allow equipment compatibility with a variety of different stainers
  • Can be loaded in mix-or-match fashion
  • Sold in sets of five
Specifications
Specifications
Coverslipper Basket/Hanger
Epredia ClearVue Coverslipper
Gemini-compatible basket, white sliders
5 Pk.