Learn More
Epredia™ Slide and Block Cabinets, Block Bank, Gray
Form an integrated storage system with stackable Epredia™ Slide and Block Cabinets, constructed of steel and finished in solvent-proof paint.
Brand:  Epredia™ 161301
 View more versions of this product
Product Code. 12553587
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Features
  • Steel construction
  • Solvent-proof paint
  • Sealed top and bottom to ensure dust-free storage
  • Universal plinth in black metal for Slide Safe and Block Bank
  • Optional Wheeled Plinth capable of supporting up to ten Slide Safes
Slide Safe
  • Contains fourteen drawers
  • Total capacity of 6000 microscope slides
  • Dimensions: 18.9 × 18.9 × 5.1" (480 × 480 × 130mm)
Block Bank
  • Contains eight drawers
  • Approximate total capacity of 3000 blocks
  • Dimensions: 18.9 × 18.9 × 10.8" (480 × 480 × 275mm)
Ordering Information:
Foam dividers for slide drawers (Cat. No. B1000152) available in packs of 12.
Order Info
Note: For a complete set, order a plinth (universal or wheeled), a block bank (choice of colors) and a slide safe (choice of colors).
Specifications
Specifications
Slide Safe
6000 Slides
480 mm
480 mm
130 mm
Blue
18.9 in.
18.9 in.
5.1 in.