missing translation for 'onlineSavingsMsg'
Learn More

Kartell Slide Storing System

Slide storing system designed for the classification and storage of microscope slides, made of high impact PMMA. The semi-transparent ice color gives an attractive appearance while the metal structure makes it stable, while ensuring the value of lightness, functionality, strength and durability.

£701.00

Specifications

Type Slide Storage Cabinet
Slide Capacity 5000
For Use With (Equipment) Classification and storage of microscope slides
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
18087761
View Documents
Kartell
00945L1
4 Swivel Castors
£701.00
Each
Please to purchase this item. Need a web account? Register with us today!
18007801
View Documents
Kartell
00946L1
4 Fixed Feet
£701.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Each single drawer can hold up to 13 supports for slides (code 921) and transparent cover plates (code 920), to be ordered separately
  • Each slides support can contain 100 microscope slides, easily extractable
  • Each cavity is numbered
  • The transparent lids guarantee an easy check of the contents, protecting the slides from dust and other foreign bodies
  • The complete Storing System can therefore contain up to 5,200 slide
Specifications

Specifications

Slide Storage Cabinet
5000
Classification and storage of microscope slides
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.