missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LTE Scientific™ Shelf and Support Brackets for 100L Drying Cabinet
Shelf and support brackets for use with LTE Scientific 100L Stainless Steel Drying Cabinet.
Brand: LTE Scientific™ 322/0101/10
Additional Details : Weight : 5.00000kg
Specifications
Shelf and Support Brackets | |
Drying Cabinet (100 L) |