missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Racks for Thermo Scientific™ 4 Shelf TSX/TDE Series Freezers

Maximize tube storage capacity per freezer with a racking solution for four inner-door, ultra-low temperature freezers.

Brand:  Thermo Scientific™ 1950520

Code : 51

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11870543

  • £224.59 / Each
In Stock
to see stock.
Add to basket

Description

Description

Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 4 shelf Series Freezers as well as well as previous generation Forma™ 88000 and TSU Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.

  • With sliding drawer racks
  • Adjustable side access box racks
  • Standard/deepwell microplate racks
Specifications

Specifications

Side Access Freezer Rack (Adjustable)
Silver
Stainless Steel
5
13.5 cm
29.5 cm
26.8 in.
2 in. Box Racks - Adjustable side access rack
Boxes (2 in.)
25 Boxes/Rack
Thermo Scientific TDE Series and TSX Series (4 shelves)
5.3 in.
11.6 in.
68.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers