missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Micronova™ SlimLine™ Buckets with Casters
SlimLine™ narrow polypropylene buckets are for use with the SlimLine™ or Slim-T™ cleanroom cart system. Available in blue, red, green or yellow.
£147.00 - £239.00
Specifications
Product Type | SlimLine Bucket |
---|---|
Material | Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel |
Autoclavable | Yes |
Capacity (English) | 6 gal. |
Care Instructions | Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage. |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
12871994
|
Micronova™
B-7DBAC |
Dark Blue |
£239.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
12812004
|
Micronova™
B-7REAC |
Red |
£147.00
Each |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||||
12832004
|
Micronova™
B-7YEAC |
Yellow |
£239.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
For use with SlimLine™ Cleanroom Cart System
- For use with SlimLine or SlimT Cleanroom Cart System
- 6 gallon capacity
- Autoclavable
- Available in blue, red, green or yellow
Specifications
SlimLine Bucket | |
Yes | |
Store at Ambient Temperature. To Autoclave - do not stack - invert Bucket to allow drainage. | |
SlimLine or SlimT Cleanroom Cart System |
Polypropylene Bucket, Nickel-plated Caster, Thermoplastic Rubber Wheel | |
6 gal. | |
Cleaning Walls, Floors and Ceilings | |
Non-sterile |