missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Slimline Flat White Light Panel
GREENER_CHOICE

Slimline Flat White Light Panel is ideal for visualising visible proteins, coomassie blue, etc.

Brand:  Syngene™ WLP-2014

328.40 GBP valid until 2024-12-31
Use promo code "21805" to get your promotional price.


Product Code. 15160587

  • £355.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with G:Box Chemi XX6 and XX9 systems
  • Size: 20 x 14 cm
  • With power supply for darkroom or benchtop use.
Specifications

Specifications

Fixed
352 nm
Syngene Gbox XX6 and XX9
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.