missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional SlimLine Lab Washer PLW 7111 S-1002

SlimLine lab washer with electric heating, hot air drying, drain pump, capacitor, water softener, oil-based app, and water protection.

Brand:  Miele Professional 11879740

Product Code. 30130357

  • £25855.00 / 1 unit

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Capacity/cycle 192 narrow neck tubes or 98 pipettes - 241 L usable volume
  • Simple and flexible - EasyLoad with flexible level positions
  • Efficient - strong pump power with the variable-speed pump
  • High capacity - reprocessing of laboratory flasks up to 50 L
Specifications

Specifications

Full-color Touch Screen
Laboratory Glassware Washer
Gray
Electrical Heating, Hot Air Drying, Drain Pump, Capacitor, Water Softener, Oil-based App, Water Protection
702 mm
650 mm
3N AC 380/415 V 50 Hz
Stainless Steel
241 L
Laboratory Glassware Reprocessing
25 5/8 in.
1965 mm
251 kg
400 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers