missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ SLiner AG Connectite Top Hole
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092325
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent instruments
Specifications
GC Inlet Liner, ConnecTite Liner Top Hole | |
78.5 mm | |
6.3 mm |
5/Pk. | |
4 mm |