missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600

Product Code. 17168590
Click to view available options
Color:
Black
Transparent
Yellow
Width (Metric):
19 mm
25 mm
51 mm
102 mm
152 mm
305 mm
508 mm
914 mm
Unit Size:
1 roll
2 rolls
4 rolls
Each
Pack of 2
Pack of 4
This item is not returnable. View return policy

Product Code. 17168590

Brand: 3M™ Safety-Walk™ 7000042869

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W) 18.3 m x 25 mm
Finish Mineral-coated
For Use With (Application) Light to Heavy Traffic Areas
Length (English) 60 ft.
Length (Metric) 18.3 m
Packaging Quantity 4 Pk.
Width (English) 1 in.
Width (Metric) 25 mm
Adhesive Type Pressure Sensitive Adhesive
Color Transparent
Type Floor Tape
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.