Learn More
3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600
Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.
£95.15 - £1407.60
Specifications
Adhesive Type | Pressure Sensitive Adhesive |
---|---|
Finish | Mineral-coated |
For Use With (Application) | Light to Heavy Traffic Areas |
Length (Metric) | 18.3 m |
Product Code | Brand | Width (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Color | Price | Quantity & Availability | ||||
17108690
|
3M™ Safety-Walk™
7100032939 |
19 mm | Black |
£95.15
4 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17118780
|
3M™ Safety-Walk™
7100158876 |
19 mm | Transparent |
£123.00
4 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17148760
|
3M™ Safety-Walk™
7100146041 |
152 mm | Transparent |
£235.00
1 roll |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17128680
|
3M™ Safety-Walk™
7100247071 |
305 mm | Black |
£610.00
Each |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17158760
|
3M™ Safety-Walk™
7100146101 |
305 mm | Transparent |
£442.00
1 roll |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17138780
|
3M™ Safety-Walk™
7100158963 |
305 mm | Yellow |
£610.00
1 roll |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
17128790
|
3M™ Safety-Walk™
7100158973 |
508 mm | Black |
£775.00
1 roll |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17128780
|
3M™ Safety-Walk™
7100158961 |
19 mm | Yellow |
£126.00
4 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17158700
|
3M™ Safety-Walk™
7100067526 |
25 mm | Black |
£204.00
Pack of 4 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17168590
|
3M™ Safety-Walk™
7000042869 |
25 mm | Transparent |
£160.00
4 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17188600
|
3M™ Safety-Walk™
7000052247 |
25 mm | Yellow |
£204.00
4 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17118560
|
3M™ Safety-Walk™
7000002135 |
51 mm | Transparent |
£204.00
Pack of 2 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17198670
|
3M™ Safety-Walk™
7100000852 |
51 mm | Yellow |
£204.00
2 rolls |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17108680
|
3M™ Safety-Walk™
7100001946 |
102 mm | Transparent |
£204.00
1 roll |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17188680
|
3M™ Safety-Walk™
7100011625 |
152 mm | Black |
£303.00
1 roll |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||||
17158680
|
3M™ Safety-Walk™
7100139113 |
914 mm | Black |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Anti-slip tape for light to heavy shoe-traffic areas
- High quality abrasive particles to ensure long-lasting friction in any application
- Highly durable surface withstands the rigors of equipment traffic
- High performance, pressure-sensitive adhesive for easy application
- Custom shapes and sizes are also available
- NFSI High Traction Certification
Specifications
Pressure Sensitive Adhesive | |
Mineral-coated | |
Light to Heavy Traffic Areas | |
18.3 m |