Learn More
Thermo Scientific™ SLIPFREE Ferrules
Shop All Thermo Scientific Products
Click to view available options
Material:
PEEK
Vespel
Unit Size:
Each
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
- Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
- Kel-F and PEEK are offered for applications in which Vespel is not suitable
Specifications
Specifications
| Type | Sealing Ferrule |
| Description | PEEK sealing ferrule |
| For Use With (Equipment) | SLIPFREE Connectors for HPLC Columns |
| Material | PEEK |