missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLN Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £385.00
Specifications
| Antigen | SLN |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17793704
|
Invitrogen™
PA5122311 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17783704
![]() |
Invitrogen™
PA5122310 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLN, also named as Sarcolipin, belongs to the sarcolipin family. It is associated with calcium ATPase SERCA1. It inhibits SERCA pumps. SLN, a key regulator of cardiac sarco (endo)plasmic reticulum (SR) Ca(2+) ATPase, is predominantly expressed in atria and mediates beta-adrenergic responses.Specifications
| SLN | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O00631 | |
| 6588 | |
| Met1-Tyr31 (Accession O00631), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Sln |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 2310045A07Rik; Sarcolipin; SLN | |
| Sln | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title