missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Assistent™ Sloping Stem Glass Funnels

For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Karl Hecht™ Assistent™ Sloping Stem Glass Funnels are manufactured from clear soda-lime glass.

Brand:  Karl Hecht™ 44000024

 View more versions of this product

Product Code. 10247391

  • £3.93 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Slope angle of 60°
  • Available in a range of diameters
Specifications

Specifications

550mm
Glass
60°
Laboratory Applications
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers