Learn More
Sartorius™ Slotted Weighing Pan for Laboratory Balances
Weighing platform for use with Sartorius™ laboratory balances.
Brand: Sartorius™ YSP01SQP
Additional Details : Weight : 0.30000kg
Description
- For use with Secura™ and Quintix™ 0.01 mg laboratory balances
Specifications
Secura™ and Quintix™ Laboratory Balances | |
80 mm Dia. | |
Weighing Pan |