missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SlowFade™ Gold Antifade Mountant
GREENER_CHOICE

Product Code. p-9056001
Click to view available options
Quantity:
2 mL
10 mL
5 x 2 mL
Unit Size:
10mL
2mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500686

Brand: Invitrogen™ S36937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Protects fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments.

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.

Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes

SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.
For Use With (Equipment) Microscope
Product Line SlowFade
Product Type Mounting Media
Quantity 5 x 2 mL
Solution Type Anti-fade Mountant
Green Features Less hazardous, sustainable packaging
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.