missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SLU-PP-332
GREENER_CHOICE

Product Code. 18724394
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18724394

Brand: Tocris Bioscience™ 8112/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent pan-ERR agonist

SLU-PP-332 is a potent and selective pan estrogen related receptor (ERR) agonist (EC50 values are 98 nM, 230 nM and 430 nM for ERRα, ERRβ and ERRγ respectively). In a mouse model of metabolic syndrome SLU-PP-332 increases energy expenditure and fatty acid oxidation, decreases fat mass, and improves insulin sensitivity. Also increases mitochondrial respiration in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-4-Hydroxy-N'-(naphthalen-2-ylmethylene)benzohydrazide
CAS 303760-60-3
Quantity 10 mg
Target Estrogen and Related Receptor Agonists
Molecular Formula C18H14N2O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.