missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets

For the safe and approved storage of aggressive, non-flammable hazardous materials in workrooms.

Brand:  Asecos™ 3293600130608

5727.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Code : NEW

Additional Details : Weight : 195.00000kg

 View more versions of this product

Product Code. 15695677

  • £6615.46 / Each
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

  • Robust construction and longevity – cabinet body made of powder-coated sheet steel with safety elements assembled outside the storage compartment for increased protection against corrosion
  • Two-in-one – two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
  • No unauthorized use – doors lockable with profile cylinder with integration in an existing locking system possible
  • Easy alignment – adjusting aids to compensate for uneven floors
  • Recirculating air filter system, ideal for the direct installation in working areas with very low noise, only approx. 39 dB
  • With multi-stage wideband filter
  • Monitoring electronics for extraction include cool-device cable and power plug
  • Optical and acoustic alarm including potential-free alarm contact
  • Display with innovative touch panel
  • Manual check of the filter saturation
  • Eco-friendly with longer filter lifetimes and a reusable filter system
Specifications

Specifications

12 x 25 kg/11 L PP pull-out shelves with PP sumps
Storage of aggressive, non-flammable hazardous materials
Steel
350.3 kg
Light Gray
Wing
EN 14727
Safety storage cabinet
Powder-coated
615 x 1197 x 2300 mm
565 x 485 x 1865 mm
12 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers