missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SmaI (10 U/μL)

Cut at CCC^GGG sites with SmaI restriction enzyme, which performs best at 30°C in Tango buffer (Isoschizomers: TspMI, XmaCI, XmaI).

Brand:  Thermo Scientific™ ER0662

 View more versions of this product

Product Code. 11833833

This item is not returnable. View return policy

Description

Description

Lambda DNA digested with SmaI, 0.7% agarose, 3 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
Sma I
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer Tango
30°C
5 x 1200 U
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.