Learn More
Hamilton™ Small Hub Removable Needle Syringe
Used exclusively on RN CTC syringes.
Brand:  Hamilton™ 203481
Product Code. 11513024
  • £28.00 / Each
Explore more special offers
Description
Description
  • Length of 51mm
  • Point style 3 (blunt)
  • Sold in packs of 3
Specifications
Specifications
Small RN (CTC) hub
22s G
3/Pk.
3