Learn More
Smart AirTherm Heater 230V
Brand:  WPI World Precision Instrument AIRTHERM-SMT-2W
Encompass_Preferred
Product Code. 15573940
  • £6173.55 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The AirTherm SMT is a new generation of heat control designed to be used in Live Cell Imaging applications with microscopes that are fitted with a full microscope environmental chamber enclosure. The standard AirTherm SMT controls temperature and, as an option, humidity. The AirTherm SMT uses a PID control algorithm to maintain tightly controlled loops of heat and humidity environment control. With AirTherm SMT, the temperature of the sample and microscope optics can be controlled to within 0.2°C. During operation, air is drawn out of the chamber through a flexible hose, heated by the AirTherm SMT heater, humidified (if desired) and re-circulated to the chamber by the return hose.