Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A
Use this kit to for purification of immunoglobulins prior to protein A digestion.
Brand:  Thermo Scientific™ 60116-101
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15870182
  • £630.07 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein A. These kits were designed to remove the challenges often associated with the process of protein clean up by providing a simple and easy capture workflow.
The SMART Digest Bulk fractionation Protein A Kits contain beads (either non-magnetic or magnetic, the magnetic allowing automation) which target purification of immunoglobulins due to the specific and strong affinity for binding to the Fc portion of the molecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
Specifications
Specifications
SMART Digest Bulk Protein A Fractionation Kit, Magnetic, Standalone
None
Immunocapture and Sample Preparation
Immunocapture
Protein A
SMART Digest Resin, Bulk Magnetic
SMART Digest
Bulk Magnetic SMART Digest Resin