Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Brand:  Thermo Scientific™ 60115-101
 View more versions of this product
Product Code. 15830182
  • £1100.48 / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, Soluble
C-terminal side of lysine and arginine amino acids
SMART Digest Soluble Resin, Bulk Non-magnetic
SMART Digest
Bulk Non-magnetic SMART Digest Soluble Resin
Protein G
Trypsin
Immunocapture and Protein Digest Sample Preparation
Immunocapture