Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific SMART Digest Bulk Fractionation Kits: Protein G.
Brand: Thermo Scientific™ 60115-102
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Protein G. These kits remove the challenges often associated with protein cleanup by providing a simple and easy capture workflow.
SMART Digest Bulk Fractionation Kits: Protein G contain beads that target purification of immunoglobulins due to the specific and strong affinity for binding to the Fab and Fc portions of the molecule. Choose from non-magnetic beads, or magnetic beads which enable automation. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once complete, perform digestion using SMART Digest enzymes if required.
Specifications
SMART Digest Bulk Fractionation Kit, Protein G, Non-Magnetic, with SOLAμ Collection Plate, Soluble | |
C-terminal side of lysine and arginine amino acids | |
SMART Digest Soluble Resin, Bulk Non-magnetic | |
SMART Digest | |
Bulk Non-magnetic SMART Digest Soluble Resin, Collection Plate, SOLAμ SPE Plate |
Protein G | |
Trypsin | |
Immunocapture and Protein Digest Sample Preparation | |
Immunocapture |