Learn More
Thermo Scientific™ SMART Digest™ Chymotrypsin Kits
Shop All Thermo Scientific ProductsDescription
Obtain fast, simple, and highly reproducible chymotrypsin protein digestion for peptide characterization and quantitation in biopharmaceutical research.
Obtain high-quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Chymotrypsin Kits. A significant advance in sample preparation for biopharmaceutical protein research, these kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Characterize and quantitate proteins faster, easier, and quicker than ever before using SMART Digest kits. This version of the kit uses chymotrypsin, which cleaves peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan.
Easy and Simple- Significantly increased speed of sample processing
- Simple process
- Easily automate entire protocol for high-throughput processing
- Increased reproducibility over existing protocols
- Higher sensitivity
- Higher data confidence
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digestion can take up to 24 hours.
Cleaner SamplesReduction in the number of chemicals used and the immobilized chymotrypsin design helps mitagate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.
Couple with SOLAμ SPE plates for desaltingWhen used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
Specifications
Specifications
| Description | with Filter/Collection plate |
| Capture | None |
| Cleavage | Peptide bonds of aromatic residues (e.g., tyrosine, phenylalanine, tryptophan) |
| Enzyme | Chymostrypsin |
| Format | SMART Digest Resin Pre-filled into PCR Tubes |
| For Use With (Application) | Peptide Quantitation, Peptide Mapping |
| Product Line | SMART Digest |
| Type | Protein Digestion |
| Includes | SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.