missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits

Product Code. p-6911039
Click to view available options
Description:
Magnetic
Magnetic with SOLAμ SPE and collection plate
Non-magnetic
Non-magnetic with SOLAμ SPE and collection plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15688683

Brand: Thermo Scientific™ 60111101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.

Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Protein A Kits. These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The SMART Digest IA Protein A Kits target purification of immunoglobulins due to the strong affinity for binding to the Fc portion of the molecule.

Solving the challenges of low-level quantitation in complex matrices

The need to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging.

As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.

Immunocapture and Protein Digestion in a Single Well

These IA kits deliver a process that is:

  • Fast
  • Simple
  • Highly reproducible
  • Sensitive
Unique Design Reduces Workflow to Hours

In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.

TRUSTED_SUSTAINABILITY

Specifications

Description Non-magnetic
Capture Protein A
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format SMART Digest Resin, Non-magnetic
For Use With (Application) Peptide Quantitation
Product Line SMART Digest
Type Immunocapture + Protein Digestion combined on a single bead
Includes Non-magnetic SMART Digest Resin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.