Learn More
Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits
Obtain fast, simple, highly reproducible, and sensitive strepavidin immunocapture and trypsin protein digestion in a single well for your biomarker studies.
£1025.10 - £1412.03
Specifications
Capture | Streptavidin |
---|---|
Cleavage | C-terminal side of lysine and arginine amino acids |
Enzyme | Trypsin |
For Use With (Application) | Peptide Quantitation |
Product Line | SMART Digest |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15648683
|
Thermo Scientific™
60110-101 |
SMART Digest IA Kit, Streptavidin (Av) non-magnetic |
£1025.10
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15658683
|
Thermo Scientific™
60110-102 |
SMART Digest IA Kit, Av non-magnetic with SOLAμ SPE and collection plate |
£1251.23
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15668683
|
Thermo Scientific™
60110-103 |
SMART Digest IA Kit, Av magnetic with SOLAμ SPE and collection plate |
£1412.03
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15678683
|
Thermo Scientific™
60110-104 |
SMART Digest IA Kit, Av magnetic |
£1115.55
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well using Thermo Scientific™ SMART Digest™ ImmunoAffinity (IA) Streptavidin Kits. These kits were designed to remove the challenges often associated with the process of biomarker quantitation by providing a simple and easy capture-digestion workflow that takes only hours instead of overnight. The kit purifies various target biomolecules via the formation of strong streptavidin-biotin bond.
Solving the Challenges of Low-level Quantitation in Complex MatricesThe need to improve the ability to identify low-level biomarkers is forcing the analytical community to deal with extremely high levels of analyte and sample complexity, which makes quantitation challenging.
As these proteins are often present at low levels in complex biological matrices, an immunoaffinity capture step is typically employed to purify the sample and increase sensitivity. This step is often followed by protein digestion. Current processes are often time consuming, multistep, and prone to irreproducibility.
Immunocapture and Protein Digestion in a Single WellThese IA kits deliver a process that is:
- Fast
- Simple
- Highly reproducible
- Sensitive
In SMART Digest IA Kits, the immunocapture reagents and the temperature-activated, thermally stable trypsin are co-immobilized onto a resin. Following enrichment, the enzyme is activated at elevated temperatures for accelerated digestion under protein denaturing conditions, resulting in a workflow which takes hours rather than overnight.
Specifications
Streptavidin | |
Trypsin | |
SMART Digest |
C-terminal side of lysine and arginine amino acids | |
Peptide Quantitation | |
Immunocapture + Protein Digestion combined on a single bead |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.