missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Trypsin Kits

Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.

£613.05 - £1005.00

Specifications

For Use With (Application) Peptide Quantitation, Peptide Mapping
View More Specs

Products 14
Product Code Brand Description Format Includes Price Quantity & Availability  
Product Code Brand Description Format Includes Price Quantity & Availability  
15382360
View Documents Promotion Details
Thermo Scientific™
60109-101
SMART Digest Trypsin Kit, with Collection plate SMART Digest Resin Pre-filled into PCR Tubes SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate
£975.00
1 set
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
16434839
View Documents Promotion Details
Thermo Scientific™
60109-101-B-LPH
Smart digest kit with bulk resin , low pH SMART Digest Low pH Resin Pre-filled into PCR Tubes SMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
£613.05
1 set
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15994417
View Documents Promotion Details
Thermo Scientific™
60109-101-LPH
Smart digest kit with collection plate, low pH SMART Digest Low pH Resin Pre-filled into PCR Tubes SMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
£628.13
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15317298
View Documents Promotion Details
Thermo Scientific™
60109-103
SMART Digest Trypsin Kit, with SOLAμ/collection plate SMART Digest Resin Pre-filled into PCR Tubes SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, SOLAμ SPE Plate
£849.23
Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
16444839
View Documents Promotion Details
Thermo Scientific™
60109-101-MB-LPH
Smart digest kit, magnectic bead, bulk, low pH SMART Digest Low pH Resin Pre-filled into PCR Tubes SMART Digest Low pH Resin Pre-filled into 96 PCR Tubes, Collection Plate
£718.58
1 set
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15357350
View Documents Promotion Details
Thermo Scientific™
60109-102
SMART Digest Trypsin Kit, with Filter/Collection plate SMART Digest Resin Pre-filled into PCR Tubes SMART Digest Resin Pre-filled into 96 PCR Tubes, Collection Plate, Filter Plate
£743.70
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
30130374
View Documents Promotion Details
Thermo Scientific™
60113-101-LPH
Soluble smart digest kit, trypsin, low pH buffer SMART Digest Low pH Buffer SMART Digest Low pH Buffer
£708.53
1 set
Please to purchase this item. Need a web account? Register with us today!
15648663
View Documents Promotion Details
Thermo Scientific™
60109-101-B
SMART Digest Trypsin Kit, Magnetic Bulk Resin option SMART Digest Resin, Bulk Magnetic Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
£644.00
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15791377
View Documents Promotion Details
Thermo Scientific™
60109-101-MB
SMART Digest Trypsin Kit, Magnetic Bulk Resin option SMART Digest Resin, Bulk Magnetic Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
£850.00
1 set
Estimated Shipment: 18-11-2024
to see stock.
Add to basket
 
15658663
View Documents Promotion Details
Thermo Scientific™
60109-102-B
SMART Digest Trypsin Kit, Bulk Resin option with Filter/Collection plate SMART Digest Resin, Bulk Non-magnetic Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
£733.65
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15668663
View Documents Promotion Details
Thermo Scientific™
60109-102-MB
SMART Digest Trypsin Kit, Magnetic Bulk Resin option with Filter/Collection plate SMART Digest Resin, Bulk Magnetic Bulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, Filter Plate
£773.85
Each
Estimated Shipment: 17-12-2024
to see stock.
Add to basket
 
15678663
View Documents Promotion Details
Thermo Scientific™
60109-103-B
SMART Digest Trypsin Kit, Bulk Resin option with SOLAμ/collection plate SMART Digest Resin, Bulk Non-magnetic Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
£844.20
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15668693
View Documents Promotion Details
Thermo Scientific™
60113-101
SMART Digest Trypsin Kit, Soluble SMART Digest Soluble Trypsin SMART Digest Soluble Trypsin, SMART Digest Buffer, Collection Plate
£1005.00
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
15688663
View Documents Promotion Details
Thermo Scientific™
60109-103-MB
SMART Digest Trypsin Kit Magnetic Bulk Resin option with SOLAμ/collection plate SMART Digest Resin, Bulk Magnetic Bulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
£924.60
Each
Estimated Shipment: 14-11-2024
to see stock.
Add to basket
 
Description

Description

Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Trypsin Protein Digestion

SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.

Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.

Couple with SOLAμ SPE Plates for Desalting

When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

Specifications

Specifications

Peptide Quantitation, Peptide Mapping
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trypsin Kits

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : sodium azide, calcium chloride

SDS
Documents

Documents

Product Certifications