missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SMART Digest™ Pepsin Kits

Product Code. 16805961 Shop All Thermo Scientific Products
Click to view available options
Description:
Bulk Resin (No Collection Plate)
with Collection Plate
with Filter and Collection Plate
with Magnetic Bead, Bulk Option (No Collection Plate)
with SOLAμ Collection Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 16805961

Brand: Thermo Scientific™ 60109110MB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these pepsin protein digestion kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results.

Use Thermo Scientific™ SMART Digest™ Pepsin Kits in your peptide mapping and peptide quantitation workflows to obtain high quality analytical results. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Pepsin Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

How SMART Digest Pepsin Kits work
Pepsin cleaves proteins preferentially at carboxylic groups of aromatic amino acids such as phenylalanine, tryptophan and tyrosine. It will not cleave at bonds containing valine, alanine or glycine. Pepsin is often used to generate antibody fragments. Pepsin is used to generate F(ab')2 fragments by removing only that portion of the Fc domain beyond the hinge region. The resulting F(ab')2 fragments are composed of two antibody-binding Fab' fragments connected by disulfides in the hinge region.

TRUSTED_SUSTAINABILITY

Specifica

Description with Magnetic Bead, Bulk Option (No Collection Plate)
Capture None
Cleavage Hydrophobic residues in P1 and P1' positions at the carboxyl side of phenylalanine and leucine and at the carboxyl side of glutamic acid
Enzyme Pepsin
Format Bulk Magnetic
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Digest Pepsin Kit
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Buffer
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato