Learn More
Thermo Scientific™ SMART Digest™ Proteinase K Kits
Obtain fast, simple, highly reproducible, and sensitive immunocapture and protein digestion in a single well for your biomarker studies.
Brand:  Thermo Scientific™ 60109-107-B
728.17 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15668673
  • £733.65 / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
SMART Digest Proteinase K Kit, Bulk Resin option
Peptide bond adjacent to carboxyl group of aliphatic and aromatic amino acids with blocked alpha amino groups
SMART Digest Resin, Bulk Non-magnetic
SMART Digest
Bulk Non-magnetic SMART Digest Resin, SMART Digest Buffer
None
Proteinase K
Peptide Quantitation, Peptide Mapping
Protein Digestion
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ SMART Digest™ Proteinase K Kits >