Learn More
Moldex™ Smart FFP-Mask
Click to view available options
For Use With (Application):
For Protection Against Fine Toxic Dust, Oil & Water Based Mists and Fumes
Nuisance Relief or Protection Against Ozone
Unit Size:
Pack of 10
Pack of 20
Description
Moldex Smart Special FFP2 NR D + Ozone 10 x WEL valved dust mask.
The activated layer of charcoal built into this mask offers relief from low levels of gases and vapours, often found in conjunction with particulate hazards. FFP2 NR D + Ozone 10 x WEL.
The activated layer of charcoal built into this mask offers relief from low levels of gases and vapours, often found in conjunction with particulate hazards. FFP2 NR D + Ozone 10 x WEL.
Specifications
Specifications
| Certifications/Compliance | EN 149:2001+A1:2009 |
| Disposable | Single-use |
| Product Type | Smart FFP-Mask |
| For Use With (Application) | Nuisance Relief or Protection Against Ozone |
| Includes | Comfortable Nose Seal |
| Quantity | 10/Pk. |