Learn More
Thermo Scientific™ SMART SPME Fibers for TriPlus™ RSH SMART Autosamplers
Track part and lot numbers, usage range, history, and more for SMART SPME Fibers on the TriPlus™ RSH SMART Autosampler.
£272.22 - £1281.04
Specifications
Length (Metric) | Phase Length: 10 mm |
---|---|
Type | Fiber |
Diameter (Metric) Outer | 23 Gauge |
Product Code | Brand | Description | Color | Phase | Thickness (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Color | Phase | Thickness (Metric) | Price | Quantity & Availability | ||
17797454
|
Thermo Scientific™
36SP01P1-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 7 μm, Green, 1/Pk. | Green | PDMS (Polydimethylsiloxane), 7 μm | 7 μm |
£272.22
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17707464
|
Thermo Scientific™
36SP01P3-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 7 μm, Green, 3/Pk. | Green | PDMS (Polydimethylsiloxane), 7 μm | 7 μm |
£674.85
Pack of 3 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
17717464
|
Thermo Scientific™
36SP03P1-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 30 μm, Golden, 1/Pk. | Golden | PDMS (Polydimethylsiloxane), 30 μm | 30 μm |
£272.22
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
17727464
|
Thermo Scientific™
36SP03P3-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 30 μm, Golden, 3/Pk. | Golden | PDMS (Polydimethylsiloxane), 30 μm | 30 μm |
£674.85
Pack of 3 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
17737464
|
Thermo Scientific™
36SP05T1-SM |
SMART SPME Fiber: DVB/Carbon WR/PDMS SMART SPME Fiber, 50/30 μm, Dark Gray, 1/Pk. | Dark Gray | DVB/Carbon WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane), 50/30 μm | 50/30 μm |
£272.22
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
17747464
|
Thermo Scientific™
36SP05T3-SM |
SMART SPME Fiber: DVB/Carbon WR/PDMS SMART SPME Fiber, 50/30 μm, Dark Gray, 3/Pk. | Dark Gray | DVB/Carbon WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane), 50/30 μm | 50/30 μm |
£674.85
Pack of 3 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
17757464
|
Thermo Scientific™
36SP06E1-SM |
SMART SPME Fiber: DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm, Violet, 1/Pk. | Violet | DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm | 65 μm |
£272.22
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
17767464
|
Thermo Scientific™
36SP06E3-SM |
SMART SPME Fiber: DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm, Violet, 3/Pk. | Violet | DVB/PDMS (Divinylbenzene/Polydimethylsiloxane), 65 μm | 65 μm |
£674.85
Pack of 3 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||
17777464
|
Thermo Scientific™
36SP08A1-SM |
SMART SPME Fiber: Acrylate (Polyacrylate), 85 μm, Gray, 1/Pk. | Gray | Acrylate (Polyacrylate), 85 μm | 85 μm |
£272.22
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
17787464
|
Thermo Scientific™
36SP08A3-SM |
SMART SPME Fiber: Acrylate (Polyacrylate), 85 μm, Gray, 3/Pk. | Gray | Acrylate (Polyacrylate), 85 μm | 85 μm |
£674.85
Pack of 3 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||
17797464
|
Thermo Scientific™
36SP08M5-SM |
SMART SPME Fibers: Collection of Five Different SMART SPME Fibers | Red/Violet/Dark Blue/Grey/Dark Grey | PDMS Thickness: 100 μmDVB/PDMS Thickness: 65 μmCarbon WR/PDMS Thickness: 95 μmAcrylate Thickness: 85 μmDVB/Carbon WR/PDMS Thickness: 50/30 μm | Various |
£1281.04
Pack of 5 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||
17707474
|
Thermo Scientific™
36SP09C1-SM |
SMART SPME Fiber: Carbon WR/PDMS (Carbon Wide Range), 95 μm, Dark Blue, 1/Pk. | Dark Blue | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 95 μm | 95 μm |
£272.22
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||
17717474
|
Thermo Scientific™
36SP09C3-SM |
SMART SPME Fiber: Carbon WR/PDMS (Carbon Wide Range) , 95 μm, Dark Blue, 3/Pk. | Dark Blue | Carbon WR/PDMS (Carbon Wide Range/ Polydimethylsiloxane), 95 μm | 95 μm |
£674.85
Pack of 3 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
17727474
|
Thermo Scientific™
36SP10P1-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 100 μm, Red, 1/Pk. | Red | PDMS (Polydimethylsiloxane), 100 μm | 100 μm |
£272.22
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
17737474
|
Thermo Scientific™
36SP10P3-SM |
SMART SPME Fiber: PDMS (Polydimethylsiloxane), 100 μm, Red, 3/Pk. | Red | PDMS (Polydimethylsiloxane), 100 μm | 100 μm |
£674.85
Pack of 3 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
Description
Track information such as part number, lot number, usage range, and history for Thermo Scientific™ SMART SPME Fibers installed on TriPlus™ RSH SMART Autosamplers. Each fiber is equipped with an ID chip, which stores this usage-based information for fast, solvent-free, and automated sample preparation. When installed on the TriPlus RSH SMART Autosampler, the information for SPME Fibers can be directly accessed and reported via Thermo Scientific™ Chromeleon™ CDS software.
Automate solvent-free solid phase micro extraction (SPME) sample preparation by performing sample extraction and enrichment in a single step with our SMART SPME Fibers.
SMART SPME Fibers also offer the additional benefit of tracking usage information. SMART SPME Fibers are compatible with all TriPlus™ RSH instruments as well as PAL platforms and AOC-6000 series instruments.
SMART Technology Benefits
- Gain higher confidence in analytical results
- Ensure error free usage of SPME Fibers through automatic identification
- Support GLP accreditation with usage history traceability
- Optimize consumption by monitoring of usage conditions and lifetime
- Facilitate SPME Fibers management thanks to color-coded health notification dashboard integrated in Chromeleon™ CDS
Specifications
Phase Length: 10 mm | |
Fiber | |
23 Gauge |