missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Smart2Pure 12 Water Purification System RO Membrane with Integrated Pretreatment
Brand: Thermo Scientific™ 09.2012
422.68 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
- RO membrane with integrated pretreatment
- For use with Smart2Pure water purification system
- 12L/hr flow rate

Specifications
Reverse Osmosis Membrane | |
12 L/hr. | |
Replacement part for Smart2Pure Water Purification System | |
12 L/hr. |
Reverse osmosis with integrated pretreatment | |
Type 3 | |
AA, IC, ICP, standard buffer |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Water analysis
File size: 7.3 MB
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Smart2Pure 12 Water Purification System RO Membrane with Integrated Pretreatment >
Spot an opportunity for improvement?Share a Content Correction