Learn More
Liebherr SmartCoolingHub
Up to 20 connectible Liebherr appliances can be linked via cable
Brand: Liebherr 6125219-00
Additional Details : Weight : 1.50000kg
Description
- Up to 48 hours of data memory if the internet connection is interrupted
- Space-saving design, wall mounting possible
- Status LED indicates operating condition
- Including 36-month licence for SmartMonitoring (valid for 1 appliance)
Specifications
RS 485 Serial Interface | |
WLAN 2.4 GHz or LAN 100Mbit |