Learn More
LevGo™ SmartSlicer™ Safety Razor Blades
Say no to dangerous sharps. SmartSlicer™ Safety Razor Blades won't scratch surfaces when cutting agarose gels.
Brand:  LevGo™ HS120259
 View more versions of this product
Product Code. 15316558
  • £44.50 / Pack of 100
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cleanly slice agarose gels* right on the light box without scratching the surface.
  • Ideal for cutting soft tissue and plant matter.
  • Adjust the size required by simply cutting with scissors.
  • Available in two sizes and made in the USA from food grade PET.
  • Supplied in packs of 100.
Specifications
Specifications
Food Grade PET
Slicing Agarose Gels, Cutting Soft Tissue and Plant Matter
Single-use
0.22 mm
100 Pk.
Blade
Small
Non-sterile
Food Grade
No