missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMEK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20348-1-AP-20UL
This item is not returnable.
View return policy
Description
Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
SMEK2 | |
Polyclonal | |
Unconjugated | |
PPP4R3B | |
FLFL2, KIAA1387, PP4R3B, PPP4R3B, PSY2, SMEK homolog 2, SMEK2, smk1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104570, 360993, 57223 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5MIZ7, Q922R5 | |
Ppp4r3b | |
SMEK2 Fusion Protein Ag14156 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |