missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMEK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SMEK2 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879874
|
Proteintech
20348-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869874
|
Proteintech
20348-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.Specifications
SMEK2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q5MIZ7, Q922R5 | |
104570, 360993, 57223 | |
SMEK2 Fusion Protein Ag14156 | |
Primary | |
-20°C | |
PPP4R3B |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FLFL2, KIAA1387, PP4R3B, PPP4R3B, PSY2, SMEK homolog 2, SMEK2, smk1 | |
Ppp4r3b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |