Learn More
ICU Medical Jelco™ Saf-T Holder™ Blood Culture Holders and Transfer Devices
Beskrivning
Saf-T Holder™ devices come with the back-end needle and multi-sample Luer permanently attached to the holder to minimize potential exposures and holder re-use. Available separately or with Saf-T Wing™ Blood Collection & Infusion Sets, Saf-T Holder™ devices also provide easy needless transfer after a syringe draw.
- Eliminate potential exposures and holder reuse
- Provide safe needleless transfer after a syringe draw
Specifikationer
Specifikationer
| For Use With (Application) | Multi-sample Luer permanently attached to the holder, provide easy needless transfer after a syringe draw |
| Includes | Female Luer |
| Product Type | Blood Culture Device |
| Model | Saf-T Holder |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.