missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Smooth Muscle Growth Supplement (SMGS)

Product Code. 10114733
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10114733

Brand: Gibco™ S00725

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Smooth Muscle Growth Supplement (SMGS)

A sterile, concentrated (20X) solution intended for use with Medium 231 to culture human smooth muscle cells. Each 25 ml bottle of SMGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human smooth muscle cells, and is the correct amount of supplement for a 500 ml bottle of basal Medium 231.

When a 500 ml bottle of medium is supplemented with SMGS, the final concentrations of the components in the supplemented medium are:

  • Fetal bovine serum (4.9% v/v)
  • Human basic fibroblast growth factor (2 ng/ml)
  • Human epidermal growth factor (0.5 ng/ml)
  • Heparin (5 ng/ml)
  • Recombinant human insulin-like growth factor-I (2μg/ml)
  • BSA (0.2μg/ml)
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -20°C).
    Cell Type Smooth Muscle Cells
    Concentration 20 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 25 mL
    Shipping Condition Dry Ice
    Species Human

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.