Learn More
Velp Scientifica™ SMS Scrubber
Designed to neutralize corrosive and toxic fumes produced during oxidative mineralizations or similar processes. Velp Scientifica™ SMS Scrubber is a space-saving and convenient solution for reducing airspace toxicity.
Brand: Velp Scientifica™ F307C0199
Code : A0
Additional Details : Weight : 3.50000kg
Description
- Complete System
- Condensation in a water trap
- Neutralization with acids or bases
- Optional absorption with activated carbon
- Ideal for high fume levels such as with soybean analysis
- Convenient to Use
- Condensate flasks removable for ease of emptying
- Easy to replace the neutralizing reagents
- Small footprint
Specifications
SMS Scrubber | |
190 x 300 x 500mm | |
DK and DKL Digestors | |
Epoxy painted stainless steel |