missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMYD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA551142
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total SMYD4 protein.
The specific function of the protein remains unknown.Specifications
SMYD4 | |
Polyclonal | |
Unconjugated | |
SMYD4 | |
G430029E23Rik; Kiaa1936; SET and MYND domain containing 4; SET and MYND domain-containing protein 4; Smyd4; ZMYND21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114826 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IYR2 | |
SMYD4 | |
Fusion protein of human SMYD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |