missing translation for 'onlineSavingsMsg'
Learn More

sn-Glycero-3-phosphocholine, MedChemExpress

MedChemExpress sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.

Brand:  Medchem Express HY-17552

Encompass_Preferred

 View more versions of this product

Product Code. 16401699

  • £68.97 / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

sn-Glycero-3-phosphocholine (Choline Alfoscerate) is a precursor in the biosynthesis of brain phospholipids and increases the bioavailability of choline in nervous tissue. sn-Glycero-3-phosphocholine (Choline Alfoscerate) has significant effects on cognitive function with a good safety profile and tolerability, and is effective in the treatment of Alzheimer's disease and dementia.
Specifications

Specifications

sn-Glycero-3-phosphocholine
White
C8H20NO6P
Choline Alfoscerate Alpha-GPC L-α-GPC
O=P(OCC[N+](C)(C)C)(OC[C@H](O)CO)[O-]
257.22
Research
Neuroscience-Neurodegeneration
28319-77-9
Research
1 g
H2O : 500 mg/mL (1943.86 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (388.77 mM; Need ultrasonic)
257.22
98.0%
Solid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research purposes only