Learn More
Thermo Scientific™ Snap-Cap Centrifuge Tubes, Plain
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
Brand: Thermo Scientific™ 3440
This item is not returnable.
View return policy
Description
Thermo Scientific™ Plain Snap-Cap Centrifuge Tubes include a flat cap, and have no frosting or graduations.
- With flat cap
- No frosting or graduations
- Certified RNase/DNase, RNA/DNA, ATP and pyrogen free
Specifications
Flat | |
25,000 x g | |
0.6 mL | |
Polypropylene | |
Non-sterile |
Certified free of RNase, DNase, DNA and pyrogens | |
Plain, Snap-Cap Centrifuge Tube | |
Natural | |
10 bags of 100 | |
No |