missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

Brand:  Eppendorf™ 0030123603

43.60 GBP valid until 2024-06-30
Use promo code "22546" to get your promotional price.



Additional Details : Weight : 405.00000kg

 View more versions of this product

Product Code. 15615367

  • £54.50 / Pack of 500
In Stock
to see stock.
Add to basket

Description

Description

  • The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
  • g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precise lid sealing minimizes evaporation rates during long-term storage.
  • The large frosted lid and side surface facilitate easy labeling.
  • Ensured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121°C, 20 min.) for effective sterilization.
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.
Specifications

Specifications

Locking Snap-cap Microtube
Hinged lid
Polypropylene
0.5 mL
Polypropylene
500 Pk.
-122.8°F to +212°F
Non-sterile
Yes
Safe-lock Lid
30,000 x g
Colorless
Autoclavable
Clear
Incubation and storage
500 Tubes (10 Bags x 50 Tubes)
-86°C to +100°C
Forensic DNA Grade (Non-sterile)
0.02 oz.
Cylindrical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers