Learn More
Fisherbrand™ Snap Cap Vial, Clear Glass
13.97 GBP valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
- A handy and inexpensive vial for uncritical analyses
- Clear
- Glass
- Flat Bottom

Specifications
Clear | |
Glass | |
Shell Vial | |
0.5 mL | |
Flat Bottom | |
100 | |
ND18 |
22 mm | |
14 mL | |
CleanPack, Polypropylene Box | |
12.7 mL | |
50 mm | |
3rd Hydrolytic Class | |
No |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products PromotionPromo Code: 24142Expires: 2025-03-14
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
47Hours
2Minutes
46Seconds
16 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
17Hours
2Minutes
46Seconds
16 -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
47Hours
2Minutes
46Seconds
16
Your input is important to us. Please complete this form to provide feedback related to the content on this product.