missing translation for 'onlineSavingsMsg'
Learn More

SNC 162, Tocris Bioscience™

Product Code. 18094462
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective non-peptide δ agonist

Potent and selective non-peptide δ-opioid receptor agonist (Ki = 0.63 nM). Displays > 8000-fold selectivity over μ-opioid receptors and is centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 178803-51-5
Molecular Formula C27H37N3O
Molecular Weight (g/mol) 419.613
InChI Key WGIDFDFAOQVAHY-UFPGJGBJSA-N
Synonym tocris-1529, 4-s-2s,5r-2,5-dimethyl-4-2-propenyl-1-piperazinyl phenylmethyl-n,n-diethylbenzamide, n,n-diethyl-4-s-alpha-2s-2beta,5alpha-dimethyl-4-allylpiperazino benzyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethylbenzamide, 4-s-2s,5r-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl-phenylmethyl-n,n-diethylbenzamide
PubChem CID 6604878
IUPAC Name 4-[(S)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-phenylmethyl]-N,N-diethylbenzamide
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC=CC=C2)N3CC(N(CC3C)CC=C)C

Specifications

Quantity 10 mg
Formula Weight 419.61
Percent Purity >99%
Chemical Name or Material SNC 162
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'