Learn More
DWK Life Sciences Sockets for Soda-lime Glass Spirit Lamps
The spirit lamp is an alternative to the gas-powered Bunsen burner and is also suitable for heating cooking glasses and the materials they contain.
Brand: DWK Life Sciences 294030008
Code : A0
Additional Details : Weight : 0.09000kg
Description
In order for substances to react with each other, a low heat input is often required. The spirit burner is used classically and in areas where direct gas extraction is not possible. It is an alternative to the well-known Bunsen burner.
- Without filling tubulature
- The ground cap ensures a tight seal and quick suffocation of the flame
Specifications
Aluminum | |
Sockets for spirit lamps (of aluminum) | |
Heating; Use with Spritus | |
50/Pk. |