Learn More
SOCS3 Mouse anti-Human, Unconjugated, Clone: 1G2B12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66797-1-IG-20UL
This item is not returnable.
View return policy
Description
The SOCS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOCS3. This antibody recognizes human, mouse antigen. The SOCS3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SOCS3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
1G2B12 | |
Unconjugated | |
20 μL | |
9021 | |
IgG1 |